, powder -600 mesh
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
93-5401-1000G
93-5401-250G
100199-596EA
151.45
USD
100199-596
100196-730
, powder -600 mesh
Molecular sieve 3A (0.3 nm, 3 Å)
Molecular Sieve
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...