Telluric acid dihydrate ≥99.5% (Te basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Hydrogen tellurate

93-5203-25G 93-5203-5G
100200-774EA 234.95 USD
100200-774 100209-148
Telluric acid dihydrate ≥99.5% (Te basis)
Telluric acid dihydrate
Formula: H₂O₄Te.2H₂0
Boiling Pt: 160 °C (dec.)
Melting Pt: 136 °C
Density: 3.071
MDL Number: MFCD00150387
CAS Number: 13451-14-4
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR