Silver tungsten oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Silver tungstate

93-4737-10G 93-4737-50G
100203-478EA 114.65 USD
100203-478 100203-480
Silver tungsten oxide ≥99%
Silver tungsten oxide
Formula: Ag₂O₄W
MDL Number: MFCD00014147
CAS Number: 13465-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR