Hexaamminenickel(II) iodide ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Hexaamminenickel(II) iodide
93-2824-25G
93-2824-100G
100196-582EA
66.52
USD
100196-582
100205-482
Hexaamminenickel(II) iodide ≥98%
Hexaamminenickel (II) iodide
Formula:
H₁₈IN₆Ni Density: 2,101 |
MDL Number:
MFCD04039936 CAS Number: 13859-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...