Iron ≥99.95%, chips

Supplier: STREM CHEMICALS INC MS

93-2664-1000G 93-2664-250G
100198-348EA 205.23 USD
100198-348 100197-776
Iron ≥99.95%, chips
Iron
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86
MDL Number: MFCD00010999
CAS Number: 7439-89-6
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR