Iron(II) chloride tetrahydrate ≥99%
Supplier: 0000018203
|
Danger
|
Synonyms:
Ferrous chloride tetrahydrate
, Iron dichloride tetrahydrate
93-2632-250G
93-2632-1000G
100209-056EA
67.94
USD
100209-056
100210-294
Iron(II) chloride tetrahydrate ≥99%
Iron (II) chloride tetrahydrate
Formula:
FeCl₂·4H₂O MW: 198.81 g/mol Melting Pt: 110 °C (dec.) Density: 1.93 |
MDL Number:
MFCD00149709 CAS Number: 13478-10-9 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...