Vanadium ≥99.9%, slug

Supplier: STREM CHEMICALS INC MS

93-2343-25G 93-2343-5G
100201-082EA 677.97 USD
100201-082 100205-432
Vanadium ≥99.9%, slug
Vanadium
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.11
MDL Number: MFCD00011453
CAS Number: 7440-62-2
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR