Disodium sulfide, anhydrous ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium sulfide, Sodium sulphide
93-1182-5G
93-1182-25G
101419-160EA
108.98
USD
101419-160
101419-158
Disodium sulfide, anhydrous ≥95%
Disodium sulphide
Formula:
Na₂S Melting Pt: 950 °C Density: 1.856 |
MDL Number:
MFCD00003498 CAS Number: 1313-82-2 UN: 1385 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...