Boranepyridine complex 93

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Pyridine Borane, Borane, pyridine complex

93-0535-10G 93-0535-50G
100196-434EA 67.94 USD
100196-434 100198-284
Boranepyridine complex 93
Boranepyridine complex
Formula: C₅H₈BN
MW: 92.94 g/mol
Boiling Pt: 65 °C (1 mmHg)
Melting Pt: 10…11 °C
Density: 0.92
Flash Pt: 70 °F
MDL Number: MFCD00012435
CAS Number: 110-51-0
UN: 1992
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR