Thallium(I) hexafluorophosphate ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Thallium(I) hexafluorophosphate(V)

81-3000-5G 81-3000-25G
100198-268EA 212.31 USD
100198-268 100198-266
Thallium(I) hexafluorophosphate ≥97%
Thallium (I) hexafluorophosphate
Formula: F₆PTl
MDL Number: MFCD00049807
CAS Number: 60969-19-9
UN: 1707
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR