Gold, nanoparticles (water soluble) [AuNP: 3nm (Citrate)]
Supplier: STREM CHEMICALS INC MS
|
Warning
|
79-0110-100ML
79-0110-25ML
101442-288EA
867.63
USD
101442-288
101442-290
Gold, nanoparticles (water soluble) [AuNP: 3nm (Citrate)]
Gold
Nanomaterials
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...