Platinum 10% on activated carbon

Supplier: STREM CHEMICALS INC MS
Danger

78-1610-25G 78-1610-5G
100202-774EA 1044.55 USD
100202-774 100199-450
Platinum 10% on activated carbon
Platinum
Formula: Pt
Boiling Pt: 3827 °C (Pt)
Density: 21.45
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR