Diammineplatinum(II) nitrite (5,0 % Pd) (w/w) in ammonium hydroxide

Supplier: STREM CHEMICALS INC MS
Danger

78-0295-10G 78-0295-50G
100198-882EA 491.14 USD
100198-882 100198-884
Diammineplatinum(II) nitrite (5,0 % Pd) (w/w) in ammonium hydroxide
Diammineplatinum(II) nitrite
Formula: H₆N₄O₄Pt
MDL Number: MFCD00011622
CAS Number: 14286-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR