Platinum ≥97% 2-5 nm

Supplier: STREM CHEMICALS INC MS
Danger

78-0007-250MG 78-0007-1G
100200-304EA 992.18 USD
100200-304 100209-270
Platinum ≥97% 2-5 nm
Platinum

Nanomaterials

Formula: Pt
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 1383
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR