Ytterbium(III) isopropoxide ≥99.9% (Yb basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Ytterbium(III) i-propoxide
70-5000-5G
70-5000-1G
100206-784EA
529.35
USD
100206-784
100203-256
Ytterbium(III) isopropoxide ≥99.9% (Yb basis)
Ytterbium (III) isopropoxide
Alkoxide
Formula:
C₉H₂₁O₃Yb Boiling Pt: 190…200 °C (0,2 mmHg) (subl.) Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00145532 CAS Number: 6742-69-4 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...