Tellurium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

52-5200-100G 52-5200-500G 52-5200-25G
100196-308EA 355.26 USD
100196-308 100195-934 100201-702
Tellurium ≥99.9%, powder
Tellurium
Formula: Te
MDL Number: MFCD00134062
CAS Number: 13494-80-9
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR