Palladium 5% on activated wood carbon wetted with ca. 50% water, paste, reduced (Escat™ 1421)

Supplier: STREM CHEMICALS INC MS
Warning

46-1903-10G 46-1903-50G
100198-840EA 559.08 USD
100198-840 100198-842
Palladium 5% on activated wood carbon wetted with ca. 50% water, paste, reduced (Escat™ 1421)
Palladium

Escat

Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR