Palladium 5% on activated wood carbon, dry, reduced (Escat™ 1431)

Supplier: STREM CHEMICALS INC MS
Danger

46-1902-10G 46-1902-50G
100198-836EA 559.08 USD
100198-836 100198-838
Palladium 5% on activated wood carbon, dry, reduced (Escat™ 1431)
Palladium

Escat

Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR