Palladium 5% on activated carbon, dried powder, reduced

Supplier: STREM CHEMICALS INC MS
Danger

46-1890-100G 46-1890-25G 46-1890-5G
100196-954EA 1620.62 USD
100196-954 100197-532 100197-534
Palladium 5% on activated carbon, dried powder, reduced
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR