Bis(tri-o-tolylphosphine)palladium(II) dichloride ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: trans-Dichlorobis(tri-o-tolylphosphine)palladium(II), Bis[tri(o-tolyl)phosphine]palladium(II) chloride, Dichlorobis(tri-o-tolylphosphine)palladium(II), Dichlorobis(tri-o-tolylphsophine)palladium

46-0525-1G 46-0525-5G
100209-972EA 60.86 USD
100209-972 100209-974
Bis(tri-o-tolylphosphine)palladium(II) dichloride ≥95%
Bis(tri-o-tolylphosphine)palladium(II) dichloride
Formula: C₄₂H₄₂Cl₂P₂Pd
MW: 786.07 g/mol
MDL Number: MFCD00274659
CAS Number: 40691-33-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR