Tetrakis[(S)-(+)-(1-adamantyl)-(N-phthalimido)acetato]dirhodium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Rh2(S-PTAD)4

45-2071-250MG 45-2071-50MG
101433-242EA 593.05 USD
101433-242 101433-240
Tetrakis[(S)-(+)-(1-adamantyl)-(N-phthalimido)acetato]dirhodium(II)
Tetrakis[(S)-(+)-(1-adamantyl)-(N-phthalimido)acetato]dirhodium(II)
Formula: C₈₀H₈₀N₄O₁₆Rh₂
MW: 1559.32 g/mol
MDL Number: MFCD09839140
CAS Number: 909389-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR