Chlorobis(ethylene)rhodium(I) dimer ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(ethylene)rhodium(I) chloride dimer, [Rh(C₂H₄)₂Cl]₂, µ-Dichlorotetraethylene dirhodium(I), Dichlorotetra(ethylene)dirhodium(I), Dichlorotetrakis(ethylene)dirhodium(I)

45-0270-250MG 45-0270-1G
100196-242EA 417.54 USD
100196-242 100201-228
Chlorobis(ethylene)rhodium(I) dimer ≥99%
Chlorobis(ethylene)rhodium(I) dimer
Formula: C₈H₁₆Cl₂Rh₂
MW: 388.93 g/mol
MDL Number: MFCD00013206
CAS Number: 12081-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR