Ruthenium 5% on activated carbon wetted_50%_Water, paste, reduced (Escat™ 4401)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-4065-25G
44-4065-5G
100196-230EA
280.25
USD
100196-230
100196-232
Ruthenium 5% on activated carbon wetted_50%_Water, paste, reduced (Escat™ 4401)
Ruthenium
Escat
Formula:
Ru MW: 101.07 |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...