Ruthenium(III) nitrosyl chloride, hydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Trichloronitrosylruthenium(III)Ruthenium(III) nitrosylchloride

44-3815-5G 44-3815-1G
100198-194EA 706.28 USD
100198-194 100198-192
Ruthenium(III) nitrosyl chloride, hydrate ≥99%
Ruthenium(III) nitrosyl chloride
Formula: Ru(NO)Cl₃
MW: 237.44 g/mol
MDL Number: MFCD00049765
CAS Number: 18902-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR