Yttrium(III) 2-ethylhexanoate ≥99.9% (Y basis) (15-17% Y), Superconductor Grade
Supplier: STREM CHEMICALS INC MS
|
Warning
|
39-2400-10G
39-2400-50G
100203-110EA
828
USD
100203-110
100203-108
Yttrium(III) 2-ethylhexanoate ≥99.9% (Y basis) (15-17% Y), Superconductor Grade
Yttrium (III) 2-ethylhexanoate
Formula:
C₂₄H₄₅O₆Y MW: 518.524 g/mol |
MDL Number:
MFCD00070441 CAS Number: 103470-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...