Strontium-2-ethylhexanoate 40% in 2-ethylhexanoic acid (8 - 12% Sr)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
2-Ethylhexanoic acid strontium salt
38-2400-25G
38-2400-100G
100207-660EA
200.98
USD
100207-660
100196-198
Strontium-2-ethylhexanoate 40% in 2-ethylhexanoic acid (8 - 12% Sr)
Strontium 2-ethylhexanoate
Formula:
C₁₆H₃₀O₄Sr MW: 374.04 g/mol |
MDL Number:
MFCD00070438 CAS Number: 2457-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...