Diethylzinc 10% (w/w) in hexanes ≥95%

Supplier: STREM CHEMICALS INC MS
Danger


Packed in a Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC).

30-3029-250G 30-3029-50G
100207-650EA 452.92 USD
100207-650 100206-576
Diethylzinc 10% (w/w) in hexanes ≥95%
Diethylzinc

ALD, Metal Alkyls, Volatile Precursors for CVD

Formula: C₄H₁₀Zn
MDL Number: MFCD00009021
CAS Number: 557-20-0
UN: 1993
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR