[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), [1,1'-Bis(diphenylphosphino)ferrocene]dichloronickel(II), [1,1’-Bis(diphenylphosphino)ferrocene]dichloronickel(II)

28-0500-5G 28-0500-1G
100196-188EA 452.92 USD
100196-188 100196-186
[1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride ≥98%
[1,1'-Bis(diphenylphosphino)ferrocene]nickel (II) chloride
Formula: C₃₄H₂₈Cl₂FeNiP₂
MW: 683.98 g/mol
MDL Number: MFCD00270284
CAS Number: 67292-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR