Bis(cyclopentadienyl)cobalt ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cobaltocene, Bis(cyclopentadienyl)cobalt(II)

27-0475-1G 27-0475-25G 27-0475-5G
100209-880EA 58.03 USD
100209-880 100203-076 100203-078
Bis(cyclopentadienyl)cobalt ≥98%
Bis(cyclopentadienyl)cobalt

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Co
MW: 189.12 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00013749
CAS Number: 1277-43-6
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR