Iron(III) meso-tetraphenylporphine chloride
Supplier: STREM CHEMICALS INC MS
Synonyms:
Iron(III) Tetraphenylporphyrin chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride, meso-Tetraphenylporphyrin iron(III) chloride, iron(III) chloride TPP, 21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride
26-2880-500MG
26-2880-2G
100198-160EA
165.6
USD
100198-160
100200-910
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride
Porphine
Formula:
C₄₄H₂₈ClFeN₄ MW: 704.02 g/mol |
MDL Number:
MFCD00012152 CAS Number: 16456-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...