Scandium (III) trifluoromethanesulfonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Scandium(III) trifluoromethanesulfonate, Scandium(III) trifluoromethanesulphonate, Scandium triflate
21-2000-1G
21-2000-5G
100201-520EA
93.42
USD
100201-520
100201-522
Scandium (III) trifluoromethanesulfonate ≥98%
Scandium (III) trifluoromethanesulphonate
Metal Triflate
Formula:
C₃F₉O₉S₃Sc |
MDL Number:
MFCD00192433 CAS Number: 144026-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...