Tetrathiafulvalene ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
16-2250-1G
16-2250-250MG
100204-990EA
370.83
USD
100204-990
100202-644
Tetrathiafulvalene ≥98%
Tetrathiafulvalene
Formula:
C₆H₄S₄ Melting Pt: 113…116 °C |
MDL Number:
MFCD00005492 CAS Number: 31366-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...