Triphenylphosphine resin on poly(styrene-divinylbenzene), 20% cross-linked

Supplier: STREM CHEMICALS INC MS
Warning

15-6730-1G 15-6730-25G 15-6730-5G
100204-982EA 92 USD
100204-982 100198-016 100206-436
Triphenylphosphine resin on poly(styrene-divinylbenzene), 20% cross-linked
Triphenylphosphine

Phosphine

Formula: C₁₈H₁₅P
MDL Number: MFCD00003043
CAS Number: 603-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR