Tri(2-furyl)phosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6372-500MG
15-6372-2G
100201-498EA
63.69
USD
100201-498
100206-426
Tri(2-furyl)phosphine ≥98%
Tri(2-furyl)phosphine
Phosphine
Formula:
C₁₂H₉O₃P Boiling Pt: 136 °C (4 mmHg) Melting Pt: 63…65 °C |
MDL Number:
MFCD00151857 CAS Number: 5518-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...