Triallylphosphine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-5700-1G
15-5700-5G
100203-006EA
294.4
USD
100203-006
100196-870
Triallylphosphine ≥97%
Triallylphosphine
Phosphine
Formula:
C₉H₁₅P Boiling Pt: 69 °C (13 mmHg) Density: 0,861 |
MDL Number:
MFCD00014956 CAS Number: 16523-89-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...