Diisopropylphosphine 10% (w/w) in hexanes ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-1796-50G 15-1796-10G
100196-088EA 738.83 USD
100196-088 100200-250
Diisopropylphosphine 10% (w/w) in hexanes ≥98%
Diisopropylphosphine

Phosphine

Formula: C₆H₁₅P
MDL Number: MFCD00233814
CAS Number: 20491-53-6
UN: 1993
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR