Diisopropylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-1795-5G 15-1795-1G
101324-616EA 791.2 USD
101324-616 101324-614
Diisopropylphosphine ≥98%
Diisopropylphosphine

Phosphine

Formula: C₆H₁₅P
MDL Number: MFCD00233814
CAS Number: 20491-53-6
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR