(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1R)-1-phenylethyl]amine, dichloromethane adduct 95

Supplier: STREM CHEMICALS INC MS
Warning

15-1520-500MG 15-1520-100MG
100197-080EA 482.65 USD
100197-080 100197-078
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1R)-1-phenylethyl]amine, dichloromethane adduct 95
(S)-(+)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)bis[(1R)-1-phenylethyl]amine, dichloromethane adduct

MONOPHOS, Phosphine

Formula: C₃₆H₃₀NO₂P
MW: 539.61 g/mol
Melting Pt: 102…103 °C
MDL Number: MFCD04117687
CAS Number: 415918-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR