2-(Dicyclohexylphosphino)-2'-methylbiphenyl ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dicyclohexyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, MePhos, 2-Dicyclohexylphosphino-2'-methylbiphenyl
15-1148-500MG
15-1148-2G
100206-356EA
82.09
USD
100206-356
100198-010
2-(Dicyclohexylphosphino)-2'-methylbiphenyl ≥98%
2-(Dicyclohexylphosphino)-2'-methylbiphenyl
Buchwald Precatalysts & Ligands, Phosphine
Formula:
C₂₅H₃₃P Melting Pt: 107…110 °C |
MDL Number:
MFCD03094577 CAS Number: 251320-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...