2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: DavePhos2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl

15-1145-50G 15-1145-500MG 15-1145-10G 15-1145-2G
100198-008EA 2527.88 USD
100198-008 100198-006 100206-354 100198-004
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl

Buchwald Precatalysts & Ligands, Phosphine

Formula: C₂₆H₃₆NP
MW: 393.55 g/mol
Melting Pt: 115…119 °C
MDL Number: MFCD02183572
CAS Number: 213697-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR