Di-i-butylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

15-1030-25G 15-1030-5G
100207-982EA 833.66 USD
100207-982 100207-984
Di-i-butylphosphine ≥97%
Di-i-butylphosphine

Phosphine

Flash Pt: –1 °F
MDL Number: MFCD00269832
CAS Number: 4006-38-6
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR