(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2R,3R)-(-)-Norphos, (R,R)-NORPHOS, (2R,3R)-(-)-2,3-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene, (1R,4S,5R,6R)-5,6-Bis(diphenylphosphaneyl)bicyclo[2.2.1]hept-2-ene

15-0140-250MG 15-0140-1G
100202-946EA 261.85 USD
100202-946 100202-948
(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene ≥95%
(2R,3R)-(-)-2,3-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene

NORPHOS, Phosphine

Formula: C₃₁H₂₈P₂
MW: 462.51 g/mol
Melting Pt: 116…119 °C
MDL Number: MFCD00085365
CAS Number: 71042-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR