Bis(trimethylsilyl)acetylene ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

14-1000-10G 14-1000-50G
100196-042EA 106.15 USD
100196-042 100209-304
Bis(trimethylsilyl)acetylene ≥97%
Bis(trimethylsilyl)acetylene
Formula: C₈H₁₈Si₂
MW: 170.4 g/mol
Boiling Pt: 133…135 °C
Melting Pt: 22…23 °C
Density: 0.752
Flash Pt: 37 °F
MDL Number: MFCD00008276
CAS Number: 14630-40-1
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR