Magnesium 2-ethylhexanoate 30 - 40% in toluene

Supplier: STREM CHEMICALS INC MS
Danger

12-1260-250G 12-1260-50G
100197-362EA 883.2 USD
100197-362 100197-364
Magnesium 2-ethylhexanoate 30 - 40% in toluene
Magnesium 2-ethylhexanoate
MDL Number: MFCD00792298
CAS Number: 15602-15-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR