Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,4,8,11-Tetraazacyclotetradecane-N,N',N'',N'''-tetraacetic acid, tetraethyl ester, 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetic Acid Tetraethyl Ester

07-1930-250MG 07-1930-1G
101442-234EA 176.92 USD
101442-234 101442-238
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate ≥98%
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate
Formula: C₂₆H₄₈N₄O₈
MDL Number: MFCD09263316
CAS Number: 126320-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR