1,2-Dimethyl-3-propylimidazolium tris(trifluoromethylsulfonyl)methide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: DMPIMe

07-0470-2G
100198-070EA 573.23 USD
100198-070
1,2-Dimethyl-3-propylimidazolium tris(trifluoromethylsulfonyl)methide ≥99%
1,2-Dimethyl-3-propylimidazolium tris(trifluoromethylsulfonyl)methide

Ionic Liquids

Density: 1.52
MDL Number: MFCD03788923
CAS Number: 169051-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR