N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate

Supplier: STREM CHEMICALS INC MS

07-0422-5G 07-0422-1G
100196-904EA 525.11 USD
100196-904 100198-068
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED)
Formula: C₂₀H₂₄N₂O₆·HCl·xH₂O
MW: 424.9 g/mol
Melting Pt: 130…134 °C
MDL Number: MFCD04113603
CAS Number: 35369-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR