(+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
N,N'-((1S,2S)-Cyclohexane-1,2-diyl)dipicolinamide
, (S,S)-DACH-Pyridyl Trost Ligand
07-0341-1G
07-0341-5G
100202-862EA
155.69
USD
100202-862
100202-864
(+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) ≥98%
(+)-N,N'-(1S,2S)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide)
TROST
Formula:
C₁₈H₂₀N₄O₂ MW: 324.3800 g/mol Melting Pt: 170 °C |
MDL Number:
MFCD02684544 CAS Number: 172138-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...