(-)-N,N'-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
N,N'-(trans-Cyclohexane-1,2-diyl)dipicolinamide, (R,R)-DACH-Pyridyl Trost Ligand
07-0340-5G
07-0340-1G
100202-860EA
618.52
USD
100202-860
100202-858
(-)-N,N'-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide) ≥98%
(-)-N,N'-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide)
TROST
Formula:
C₁₈H₂₀N₄O₂ Melting Pt: 170 °C |
MDL Number:
MFCD02684543 CAS Number: 218290-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...