trans-N1,N2-Dimethylcyclohexane-1,2-diamine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: trans-N,N'-Dimethyl-1,2-cyclohexanediamine

07-0270-500MG 07-0270-2G
100197-988EA 185.42 USD
100197-988 100206-218
trans-N1,N2-Dimethylcyclohexane-1,2-diamine ≥98%
trans-N1,N2-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
MW: 142.24 g/mol
Melting Pt: 4 °C
MDL Number: MFCD03001702
CAS Number: 67579-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR